Carbenicillin Dinatriumsalz
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
CAS No.: 4800-94-6
Molecular Formula: C17H16N2Na2O6S
Formula Weight: 422.36
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Beilstein Reference: 5722128
Merck Reference: 14,1792
MDL No.: MFCD00077683
Water-soluble antibiotic effective against Gram-negative bacteria
Formel:
C₁₇H₁₆N₂O₆S·2Na Lagertemperatur: Kühlschrank |
MDL:
MFCD00077683 CAS-Nummer: 4800-94-6 EINECS: 225-360-8 Merck Index: 14,1792 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...