Benztropine mesylate

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cogentin

J61954.03
J61954.03EA 21.8 EUR
J61954.03
Benztropine mesylate
Benztropine mesylate

CAS No.: 132-17-2 Molecular Formula: C21H25NO.CH3SO3H Formula Weight: 403.54 Physical Form: Solid Appearance: White to off-white Melting Point: 135° UN No.: UN2811 MDL No.: MFCD00074784


Applications: Muscarinic receptor antagonist

Formel: C₂₂H₂₉NO₄S
Molecular Weight: 403,537 g/mol
Schmelzpunkt: 135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074784
CAS-Nummer: 132-17-2
EINECS: 205-048-8
ADR: 6.1,III
Merck Index: 14,1121

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR