Inosine 5'-monophosphate disodium salt hydrate

Lieferant: Thermo Fisher Scientific

Synonyme: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

J61959.06 J61959.14 J61959.22
J61959.06EA 32.7 EUR
J61959.06 J61959.14 J61959.22
Inosine 5'-monophosphate disodium salt hydrate
Inosine 5'-monophosphate disodium salt hydrate

CAS No.: 4691-65-0 Molecular Formula: C10H11N4Na2O8P.xH2O Formula Weight: 392.17(anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00036201

Formel: C₁₀H₁₁N₄Na₂O₈P·xH₂O
Molecular Weight: 392.17 g/mol (anhy)
MDL: MFCD00150372
CAS-Nummer: 4691-65-0
EINECS: 225-146-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR