Ciprofloxacin Hydrochlorid Monohydrat 98,0-102,0%

Lieferant: Thermo Fisher Scientific

J61970.06 J61970.14 J61970.PG J61970.A3 J61970.A1
J61970.06EA 69 EUR
J61970.06 J61970.14 J61970.PG J61970.A3 J61970.A1
Ciprofloxacin Hydrochlorid Monohydrat 98,0-102,0%
Ciprofloxacin hydrochloride monohydrate

An anti-bacterial agent effective against anaerobic bacteria


  • White/pale yellow lyophilizedpowder


CAS No.: 86393-32-0 Molecular Formula: C17H18FN3O3.HCl Formula Weight: 367.80 Storage Temperature: -30°C to -10°C Physical Form: Lyophilized powder Appearance: White/pale yellow MDL No.: MFCD00242856


Ciprofloxacin hydrochloride is used as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. Nalidixic acid analogue.

Formel: C₁₇H₁₈FN₃O₃·HCl·H₂O
Lagertemperatur: Gefrierschrank
MDL: MFCD00242856
CAS-Nummer: 86393-32-0
Merck Index: 14,2314

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to off-white
Form Lyophilised powder
Assay (unspecified) 98,0 - 102,0%
Residue on ignition 0,1% max.
Impurity content 0,002% max. (Heavy metals)
Water Content (Karl Fischer Titration) 4,7 - 6,7%
Solubility Soluble in Water (35 mg/ml)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR