Nickelsulfat 0.5 M in wässriger Lösung

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Nickel(II)sulfat

J61993.AEEA 75.1 EUR
J61993.AE
Nickelsulfat 0.5 M in wässriger Lösung
Nickelsulfat

CAS No.: 10101-97-0 Molecular Formula: NiO4S Formula Weight: 262.86 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Green Odour: Odourless UN No.: 3082 MDL No.: MFCD00011146

Formel: NiO₄S
Molecular Weight: 154,75 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011146
CAS-Nummer: 7786-81-4
EINECS: 232-104-9
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR