A-7 hydrochloride
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 79127-24-5
Molecular Formula: C20H29ClN2O2S.HCl
Formula Weight: 433.44
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Solubility: Soluble to 50mM in DMSO
MDL No.: MFCD00209822
Applications: Calmodulin antagonist
Formel:
C₂₀H₂₉ClN₂O₂S·HCl |
MDL:
MFCD00209822 CAS-Nummer: 79127-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...