Sodium pyrophosphate, 200 mM buffer solution
Lieferant: Thermo Fisher Scientific
J62052.AE
J62052.AK
J62052.AEEA
68.3
EUR
J62052.AE
J62052.AK
Sodium pyrophosphate, 200 mM buffer solution
Puffer
Klonpuffer
CAS No.: 7758-16-9
Physical Form: Liquid
MDL No.: MFCD00014246
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...