Terazosin hydrochloride ≥99%
Lieferant: Thermo Scientific
Synonyme:
(4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(tetrahydrofuran-2-yl)methanone hydrochloride
J62055.MF
J62055.MFEA
145
EUR
J62055.MF
Terazosin hydrochloride ≥99%
Terazosin hydrochloride
Purity: 99+%
CAS No.: 63074-08-8
Molecular Formula: C19H25N5O4.HCl
Formula Weight: 423.90
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00467965
Applications: Alpha-1 adrenoceptor antagonist
Formel:
C₁₉H₂₅N₅O₄·HCl Molecular Weight: 423,90 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00467965 CAS-Nummer: 63074-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...