alle-trans-Retinol (synthetisch) 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol
J62079.03
J62079.03EA
374
EUR
J62079.03
alle-trans-Retinol (synthetisch) 95%
alle-trans-Retinol
Purity: 95%
CAS No.: 68-26-8
Molecular Formula: C20H30O
Formula Weight: 286.46
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Sensitivity: Moisture and Light Sensitive
Appearance: Off-white to pale yellow
Beilstein Reference: 403040
Merck Reference: 14,10013
MDL No.: MFCD00001552
Formel:
C₂₀H₃₀O Molecular Weight: 286,46 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00001552 CAS-Nummer: 68-26-8 EINECS: 200-683-7 Merck Index: 14,10013 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...