Dextransulfat Natriumsalz, M.W. ∼ 8,000

Lieferant: Thermo Fisher Scientific

J62101.14 J62101.22 J62101.A1
J62101.14EA 73 EUR
J62101.14 J62101.22 J62101.A1
Dextransulfat Natriumsalz, M.W. ∼ 8,000
Dextran sulphate sodium salt

CAS No.: 9011-18-1 Physical Form: Powder Appearance: Off-white to pale yellow Odour: Phenol-like MDL No.: MFCD00081551

Formel: (C₆H₇Na₃O₁₄S₃)n
Lagertemperatur: Raumtemperatur
MDL: MFCD00081551
CAS-Nummer: 9011-18-1
Merck Index: 14,2951

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR