Laemmli SDS sample buffer with pyronin Y, reducing (4X)

Lieferant: Thermo Fisher Scientific

J62115.ACEA 54.3 EUR
J62115.AC J62115.AD
Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Puffer Elektrophorese-Puffer Beladungspuffer

Physical Form: Liquid
Appearance: Pink
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR