Hesperidin 95%
Lieferant: Thermo Fisher Scientific
J62126.14
J62126.22
J62126.14EA
49.3
EUR
J62126.14
J62126.22
Hesperidin 95%
Hesperidin
Purity: 95%
CAS No.: 520-26-3
Molecular Formula: C28H34O15
Formula Weight: 610.56
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to pale brown
Melting Point: 250-255° dec.
Beilstein Reference: 75140
Merck Reference: 14,4671
MDL No.: MFCD00075663
Acts as an antioxidant
Formel:
C₂₈H₃₄O₁₅ Schmelzpunkt: 250…255 °C (Zersetzung) |
MDL:
MFCD00075663 CAS-Nummer: 520-26-3 EINECS: 208-288-1 Merck Index: 14,4671 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...