bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342) 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Hoechst 33342, 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, bisBenzimide

J62134.MC
J62134.MCEA 135 EUR
J62134.MC
bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342) 98%
bisBenzimide H-33342 trihydrochloride trihydrate (Hoechst 33342)

Purity: 98% CAS No.: 23491-52-3 Molecular Formula: C27H28N6O.3HCl.3H2O Formula Weight: 615.99 (561.94anhy) Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Yellow to green Beilstein Reference: 4088183 MDL No.: MFCD00012679


Applications: Stain for identifying specific chromosome banding patterns

Formel: C₂₇H₂₈N₆O·3HCl·3H₂O
Molecular Weight: 615,99 g/mol (561,94 anhy)
Lagertemperatur: Kühlschrank
MDL: MFCD00012679
CAS-Nummer: 23491-52-3
EINECS: 245-691-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR