Zinkchlorid 25 mM in wässriger Lösung
Lieferant: Thermo Fisher Scientific
J62160.AE
J62160.AEEA
54.1
EUR
J62160.AE
Zinkchlorid 25 mM in wässriger Lösung
Zinkchlorid
CAS No.: 7646-85-7
Molecular Formula: Cl2Zn
Formula Weight: 136.28
Physical Form: Liquid
MDL No.: MFCD00011295
Formel:
ZnCl₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011295 CAS-Nummer: 7646-85-7 EINECS: 231-592-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...