5-Aminofluorescein (isomer I) 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer I), 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-Aminofluorescein, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine (isomer I)
Purity: 96%
CAS No.: 3326-34-9
Molecular Formula: C20H13NO5
Formula Weight: 347.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Dark red to brown
Melting Point: 223° dec.
Beilstein Reference: 48395
MDL No.: MFCD00005052
Applications: A fluorescent labeling reagent
Formel:
C₂₀H₁₃NO₅ Molecular Weight: 347.33 g/mol Schmelzpunkt: 223 °C (Zers.) |
MDL:
MFCD00005052 CAS-Nummer: 3326-34-9 EINECS: 222-043-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...