IEF Cathode buffer (10X) with lysine and arginine

Lieferant: Thermo Fisher Scientific

J62204.AP
J62204.APEA 88.2 EUR
J62204.AP
IEF Cathode buffer (10X) with lysine and arginine
Puffer Elektrophorese-Puffer Laufpuffer

Physical Form: Liquid
Appearance: Colourless
Applications: For isoelectric focusing

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR