Sudanschwarz B
Lieferant: Thermo Fisher Scientific
CAS No.: 4197-25-5
Molecular Formula: C29H24N6
Formula Weight: 456.55
Physical Form: Powder
Appearance: Dark brown to black
Melting Point: 118-126°
Beilstein Reference: 723248
Merck Reference: 14,8885
MDL No.: MFCD00006919
Histochemical stain for chromosomes
Formel:
C₂₉H₂₄N₆ Schmelzpunkt: 118…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006919 CAS-Nummer: 4197-25-5 EINECS: 224-087-1 Merck Index: 14,8885 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...