Triton® X-100 lysis buffer, pH 7,4

Lieferant: Thermo Fisher Scientific

J62289.AK J62289.AP
J62289.AKEA 69.9 EUR
J62289.AK J62289.AP
Triton® X-100 lysis buffer, pH 7,4
Puffer Extraktionspuffer

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR