L-Cystine disodium salt ≥98%
Lieferant: Thermo Fisher Scientific
J62310.30
J62310.A1
J62310.30EA
117
EUR
J62310.30
J62310.A1
L-Cystine disodium salt ≥98%
Purity: 98+%
CAS No.: 64704-23-0
Molecular Formula: C6H10N2Na2O4S2
Formula Weight: 284.30
Physical Form: Powder
Appearance: Pale brown
Optical Rotation: +170° (c=2 in 1N HCl)
MDL No.: MFCD00037206
Formel:
C₆H₁₀N₂Na₂O₄S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037206 CAS-Nummer: 64704-23-0 EINECS: 265-025-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...