Berberine chloride
Lieferant: Thermo Scientific
Synonyme:
9,10-Dimethoxy-5,6-dihydro-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinolin-7-ium chloride
J62311.06
J62311.06EA
45.1
EUR
J62311.06
Berberine chloride
Berberine chloride
CAS No.: 633-65-8
Molecular Formula: C20H18ClNO4
Formula Weight: 371.82
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Yellow to orange
Beilstein Reference: 3836585
MDL No.: MFCD00011939
Applications: Apoptosis inhibitor. Increases P-glycoprotein expression in hepatoma cells. Stain for heparin in mast cells
Formel:
C₂₀H₁₈ClNO₄ Molecular Weight: 371,817 g/mol |
MDL:
MFCD00011939 CAS-Nummer: 633-65-8 EINECS: 211-195-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...