Thiostrepton ≥90%

Lieferant: Thermo Fisher Scientific

Synonyme: N-[(7R,8S)-2-Carboxy-7,8-dihydro-8-hydroxy-4-[(1S)-1-hydroxyethyl]-7-quinolinyl]-L-isoleucyl-L-alanyl-2,3-didehydroalanyl-L-alanyl-2-[(4aR,11S,14Z,18S,21S,28S,32aS)-4a-amino-21-[(1S,2R)-1,2-dihydroxy-1-methylpropyl]-14-ethylidene-3,4,4a,9,10,11,12,13,14,18,19,20,21,27,28,32a-hexadecahydro-11,28-bis[(1R)-1-hydroxyethyl]-9,12,19,26-tetraoxo-17H,26H-8,5:18,15:25,22:32,29-tetranitrilo-5H,15H-pyrido[3,2-m][1,11,17,24,4,7,20,27]tetrathiatetraazacyclotriacontin-2-yl]-4-thiazolecarbonyl-2,3-didehydroalanyl-2,3-didehydro-alaninamide,(1→528)-lactone

J62332.03
J62332.03EA 180 EUR
J62332.03
Thiostrepton ≥90%
Thiostrepton

Purity: 90+% CAS No.: 1393-48-2 Molecular Formula: C72H85O18N19S5 Formula Weight: 1664.89 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to pale yellow Solubility: Soluble in DMSO or chloroform MDL No.: MFCD00135828


Applications: Natural cyclic oligopeptide antibiotic

Formel: C₇₂H₈₅N₁₉O₁₈S₅
Molecular Weight: 1664.887 g/mol
MDL: MFCD00135828
CAS-Nummer: 1393-48-2
EINECS: 215-734-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR