β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)) 97%

Lieferant: Thermo Fisher Scientific

Synonyme: NAD+, Nicotinamid-Adenin-Dinucleotid, oxidiert, Nadid, Nicotinsäureamid-Adenin-Dinucleotid, β-Nicotinamid-Adenin-Dinucleotid

J62337.03 J62337.06 J62337.14
J62337.03EA 51.2 EUR
J62337.03 J62337.06 J62337.14
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)) 97%
β-Nicotinamid-Adenin-Dinucleotid, oxidierte Form (NAD)

Purity: 98+% CAS No.: 53-84-9 Molecular Formula: C21H27N7O14P2 Formula Weight: 663.43 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Solubility: Soluble in water at 50mg/ml Beilstein Reference: 3584133 MDL No.: MFCD00036253

Formel: C₂₁H₂₇N₇O₁₄P₂
Molecular Weight: 663.43 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036253
CAS-Nummer: 53-84-9
EINECS: 200-184-4
Merck Index: 14,6344

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR