Brefeldin A ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural

J62340.MB J62340.MF
J62340.MBEA 341 EUR
J62340.MB J62340.MF
Brefeldin A ≥99%
Brefeldin A
Formel: C₁₆H₂₄O₄
Schmelzpunkt: 201…205 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00083258
CAS-Nummer: 20350-15-6
Merck Index: 14,1369

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR