Croton oil
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 8001-28-3
Flash Point: 74°(165°F)
Physical Form: Oil
Appearance: Pale yellow to brown
Odour: Pungent
MDL No.: MFCD00130880
Applications: A natural source of phorbol, phorbol esters and glycerides of saturated fatty acids
Flammpunkt:
74 °C (165 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130880 CAS-Nummer: 8001-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...