Kaliumchlorid 1 M aqueous solution RNase_free

Lieferant: Thermo Fisher Scientific

Synonyme: Potassiumchloride

J62422.AK
J62422.AKEA 76.6 EUR
J62422.AK
Kaliumchlorid 1 M aqueous solution RNase_free
Potassium chloride

CAS No.: 7447-40-7 Molecular Formula: ClK Formula Weight: 74.55 Physical Form: Liquid MDL No.: MFCD00011360

Formel: KCl
Molecular Weight: 74.55 g/mol
Siedepunkt: 1420
Schmelzpunkt: 773
Dichte: 1.13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011360
CAS-Nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR