KN-62
Lieferant: Thermo Scientific
Synonyme:
(S)-4-(2-(N-Methylisoquinoline-5-sulphonamido)-3-oxo-3-(4-phenylpiperazin-1-yl)propyl)phenyl isoquinoline-5-sulphonate
J62444.MA
J62444.MAEA
573
EUR
J62444.MA
KN-62
KN-62
Potently and specifically inhibits calcium/calmodulin kinase II.
Yellow powder
Formel:
C₃₈H₃₅N₅O₆S₂ Molecular Weight: 721,84 g/mol Schmelzpunkt: 65…67 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00083180 CAS-Nummer: 127191-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...