Melatonin ≥99%
Lieferant: Thermo Scientific
Synonyme:
Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
J62452.14
J62452.14EA
156
EUR
J62452.14
Melatonin ≥99%
Melatonin
Purity: 99+%
CAS No.: 73-31-4
Molecular Formula: C13H16N2O2
Formula Weight: 232.28
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 116-120°
Solubility: Soluble in ethanol to at least 50mg/ml
Beilstein Reference: 205542
Merck Reference: 14,5816
MDL No.: MFCD00005655
A pineal gland hormone associated with sleep regulation. It induces apoptosis
Formel:
C₁₃H₁₆N₂O₂ Molecular Weight: 232,28 g/mol Schmelzpunkt: 116…120 °C |
MDL:
MFCD00005655 CAS-Nummer: 73-31-4 EINECS: 200-797-7 Merck Index: 14,5816 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...