Fast Blue BB base
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4'-Amino-2',5'-diethoxybenzanilide, N-(4-Amino-2,5-diethoxyphenyl)benzamide, Azoic Diazo No. 20, Fast blue BB, C.I. 37175, Azoic diazo component 20 (base)
J62456.14
J62456.22
J62456.14EA
56
EUR
J62456.14
J62456.22
Fast Blue BB base
Fast Blue BB base
CAS No.: 120-00-3
Molecular Formula: C17H20N2O3
Formula Weight: 300.35
Physical Form: Powder
Appearance: Gray to pale brown
Melting Point: 95-105°
Beilstein Reference: 3416889
MDL No.: MFCD00009091
Applications: C.I. 37175
Formel:
C₁₇H₂₀N₂O₃ Schmelzpunkt: 95…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009091 CAS-Nummer: 120-00-3 EINECS: 204-363-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...