L(-)-Tryptophan Reagenz für die Zellkultur
Lieferant: Thermo Fisher Scientific
J62508.09
J62508.22
J62508.A1
J62508.09EA
35.9
EUR
J62508.09
J62508.22
J62508.A1
L(-)-Tryptophan Reagenz für die Zellkultur
L(-)-Tryptophan
CAS No.: 73-22-3
Molecular Formula: C11H12N2O2
Formula Weight: 204.23
Physical Form: Powder
Appearance: White to off-white
Melting Point: 280-285°
Beilstein Reference: 86197
Merck Reference: 14,9797
MDL No.: MFCD00064340
Formel:
C₁₁H₁₂N₂O₂ Schmelzpunkt: 280…285 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064340 CAS-Nummer: 73-22-3 EINECS: 200-795-6 Merck Index: 14,9797 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...