(S)-(+)-Camptothecin ≥90%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
Cytotoxic antitumor agent that inhibits topoisomerase I.
Formula Weight: 348.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Yellow
Melting Point: 260° dec.
Beilstein Reference: 631069
UN No.: UN1544
MDL No.: MFCD00081076
Applications: Cytotoxic antitumor agent that inhibits topoisomerase I
Formel:
C₂₀H₁₆N₂O₄ Schmelzpunkt: 260 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00081076 CAS-Nummer: 7689-03-4 UN: 1544 ADR: 6.1,III Merck Index: 14,1735 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...