TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100
Lieferant: Thermo Fisher Scientific
J62533.AP
J62533.APEA
61.9
EUR
J62533.AP
TRIS-buffered saline (TBS, 10X), with 1% Triton® X-100
Puffer
Elektrophorese-Puffer
Waschpuffer
Physical Form: Liquid
MDL No.: MFCD00132476
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...