ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2) 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, NADP-Na2, ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, Nadidphosphat Dinatriumsalz, NADP Dinatriumsalz
J62556.MD
J62556.03
J62556.MDEA
105
EUR
J62556.MD
J62556.03
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2) 97%
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Purity: 97%
CAS No.: 24292-60-2
Molecular Formula: C21H26N7Na2O17P3
Formula Weight: 787.37
Storage Temperature: -30°C to -10°C
Physical Form: Powder
MDL No.: MFCD00065390
Formel:
C₂₁H₂₆N₇Na₂O₁₇P₃ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065390 CAS-Nummer: 24292-60-2 EINECS: 246-129-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...