ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2) 97%
Lieferant: Thermo Fisher Scientific
J62556.MD
J62556.03
J62556.MDEA
105
EUR
J62556.MD
J62556.03
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2) 97%
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)
Formel:
C₂₁H₂₆N₇Na₂O₁₇P₃ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065390 CAS-Nummer: 24292-60-2 EINECS: 246-129-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...