Sorbitanmonooleat

Lieferant: Thermo Scientific

Synonyme: Sorbitanmono(cis-9-octadecenoat), Span 80, Sorbitanoleat

J62576.K2
J62576.K2EA 94.3 EUR
J62576.K2
Sorbitanmonooleat
Sorbitanmonooleat
Formel: C₂₄H₄₄O₆
Molecular Weight: 428,61 g/mol
Flammpunkt: 113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00080948
CAS-Nummer: 1338-43-8
EINECS: 215-665-4
Merck Index: 14,8724

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR