(+)-1-Deoxynojirimycin
Lieferant: Thermo Scientific
Synonyme:
1-Deoxynojirimycin, (2R,3R,4R,5S)-2-(Hydroxymethyl)piperidine-3,4,5-triol
J62602.MC
J62602.MCEA
770
EUR
J62602.MC
(+)-1-Deoxynojirimycin
(+)-1-Deoxynojirimycin
CAS No.: 19130-96-2
Molecular Formula: C6H13NO4
Formula Weight: 163.17
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 195-196°
MDL No.: MFCD00063474
Applications: An alpha-glucosidase inhibitor. Interferes with normal processing of N-linked glycoproteins.
Formel:
C₆H₁₃NO₄ Molecular Weight: 163,17 g/mol Schmelzpunkt: 195…196 °C |
MDL:
MFCD00063474 CAS-Nummer: 19130-96-2 Merck Index: 14,2905 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...