TRIS-buffered saline (TBS, 10X, high salt) pH 7,4

Lieferant: Thermo Fisher Scientific

J62664.AP J62664.K2 J62664.K3
J62664.APEA 53.6 EUR
J62664.AP J62664.K2 J62664.K3
TRIS-buffered saline (TBS, 10X, high salt) pH 7,4
Puffer Elektrophorese-Puffer Waschpuffer

CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00004679

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR