G418 Disulfat Reagenz für die Zellkultur
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
G-418 Sulfat, G418 Sulfat, Geneticin®, G-418 Disulfat
J62671.03
J62671.06
J62671.14
J62671.03EA
113
EUR
J62671.03
J62671.06
J62671.14
G418 Disulfat Reagenz für die Zellkultur
G418 disulphate
CAS No.: 108321-42-2
Molecular Formula: C20H40N4O10.2H2SO4
Formula Weight: 692.71
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 50mg/ml
MDL No.: MFCD00058314
Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths
Formel:
C₂₀H₄₄N₄O₁₈S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058314 CAS-Nummer: 108321-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...