Cefotaxim Natriumsalz
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
J62690.03
J62690.06
J62690.14
J62690.03EA
107
EUR
J62690.03
J62690.06
J62690.14
Cefotaxim Natriumsalz
Cefotaxime sodium salt
CAS No.: 64485-93-4
Molecular Formula: C16H16N5NaO7S2
Formula Weight: 477.40
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Freely soluble in water
Beilstein Reference: 5711411
MDL No.: MFCD00079073
Formel:
C₁₆H₁₇N₅O₇S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079073 CAS-Nummer: 64485-93-4 EINECS: 264-915-9 Merck Index: 14,1933 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...