Ketoprofen
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid
J62702.06
J62702.14
J62702.06EA
85.2
EUR
J62702.06
J62702.14
Ketoprofen
Ketoprofen
Formel:
C₁₆H₁₄O₃ Schmelzpunkt: 92…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055790 CAS-Nummer: 22071-15-4 EINECS: 244-759-8 UN: 2811 ADR: 6.1,III Merck Index: 14,5305 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...