TES-Natriumsalz (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure Natriumsalz)
Lieferant: Thermo Fisher Scientific
CAS No.: 70331-82-7
Molecular Formula: C6H14NNaO6S
Formula Weight: 251.23
Physical Form: Powder
Appearance: White
Beilstein Reference: 1957061
Merck Reference: 14,9176
MDL No.: MFCD00065482
A zwitterionic Good's Buffer
Formel:
C₆H₁₄NNaO₆S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065482 CAS-Nummer: 70331-82-7 Merck Index: 14,9176 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...