Orange G für die Elektrophorese

Lieferant: Thermo Fisher Scientific

Synonyme: Acid Orange 10, Dinatrium-7-hydroxy-8-phenylazonaphthalin-1,3-disulfonat, Orange G Natriumsalz, CI 16230

J62743.14 J62743.22 J62743.A1
J62743.14EA 19.7 EUR
J62743.14 J62743.22 J62743.A1
Orange G für die Elektrophorese
Orange G

CAS No.: 1936-15-8 Molecular Formula: C16H10N2Na2O7S Formula Weight: 452.38 Physical Form: Powder Appearance: Red to orange Solubility: Soluble in water or methyl cellosolve. Slightly soluble in ethanol Beilstein Reference: 4120705 MDL No.: MFCD00012457


Collagen stain for connective tissue

Formel: C₁₆H₁₀N₂Na₂O₇S₂
Schmelzpunkt: 141 °C
Dichte: 0,45…0.47 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012457
CAS-Nummer: 1936-15-8
EINECS: 217-705-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR