Dextransulfat Natriumsalz, M.W. ~ 500,000
Lieferant: Thermo Fisher Scientific
J62787.06
J62787.18
J62787.22
J62787.A1
J62787.06EA
43.7
EUR
J62787.06
J62787.18
J62787.22
J62787.A1
Dextransulfat Natriumsalz, M.W. ~ 500,000
Dextran sulphate sodium salt
CAS No.: 9011-18-1
Physical Form: Powder
Appearance: Off-white to pale yellow
Odour: Phenol-like
Solubility: Soluble in water
Merck Reference: 14,2951
MDL No.: MFCD00081551
Formel:
(C₆H₇Na₃O₁₄S₃)n Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081551 CAS-Nummer: 9011-18-1 Merck Index: 14,2951 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...