Tropisetron hydrochloride
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(3-endo)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl 1H-Indole-3-carboxylate Hydrochloride, ICS 25-93, SDZ ICS-93, 3-Tropanylindole-3-carboxylate hydrochloride, endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-yl 1H-indole-3-carboxylate hydrochloride
J62807.MC
J62807.MCEA
190
EUR
J62807.MC
Tropisetron hydrochloride
Tropisetron hydrochloride
CAS No.: 105826-92-4
Molecular Formula: C17H20N2O2.HCl
Formula Weight: 320.82
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water
UN No.: UN2811
MDL No.: MFCD00210221
Applications: A selective 5-HT3 serotonin receptor antagonist
Formel:
C₁₇H₂₀N₂O₂ Molecular Weight: 320,82 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00210221 CAS-Nummer: 105826-92-4 UN: 2811 ADR: 6.1,III Merck Index: 14,9783 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...