Gentamicinsulfat 600 IU/mg

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Gentamicin Sulfatsalz

J62834.03 J62834.06 J62834.09
J62834.03EA 38.7 EUR
J62834.03 J62834.06 J62834.09
Gentamicinsulfat 600 IU/mg
Gentamicinsulfat

Gentamycin Sulfate is a broad-spectrum, aminoglycoside antibiotic used for cell culture which inhibits protein synthesis in sensitive organisms. Reported to have activity against gram-negative bacteria, Pseudomonas aeruginosa, gram-positive Staphylococcus aureus and some mycoplasmas. Reported to have low toxicity to mammalian cells and viruses at certain concentrations, useful for long-term tissue culture studies with viruses.


Composed of Gentamycin C1, Gentamycin C1a and Gentamycin C2.


Applications: Bacterial antibiotic that inhibits protein synthesis


Bitte beachten Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Lagertemperatur: Raumtemperatur
MDL: MFCD00270181
CAS-Nummer: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR