Staurosporine ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282
Purity: 99+%
CAS No.: 62996-74-1
Molecular Formula: C28H26N4O3
Formula Weight: 466.54
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 240-243°
Solubility: Soluble in DMSO or ethanol
Beilstein Reference: 1060573
MDL No.: MFCD00077402
Applications: Broad spectrum protein kinase inhibitor
Formel:
C₂₈H₂₆N₄O₃ Molecular Weight: 466.541 g/mol Schmelzpunkt: 240…243 °C |
MDL:
MFCD18252446 CAS-Nummer: 62996-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...