Ethyl orange sodium salt
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Diethylaminoazobenzene-4'-sulphonic Acid Sodium Salt
J62863.22
J62863.22EA
33.8
EUR
J62863.22
Ethyl orange sodium salt
Ethyl orange sodium salt
CAS No.: 62758-12-7
Molecular Formula: C16H18N3NaO3S
Formula Weight: 355.39
Physical Form: Powder
Appearance: Orange
Beilstein Reference: 3821641
MDL No.: MFCD00007503
Applications: Indicator: pH 3.0 (red) to pH 4.5 (yellow)
Formel:
C₁₆H₁₈N₃NaO₃S Molecular Weight: 355,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007503 CAS-Nummer: 62758-12-7 EINECS: 263-716-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...