Thapsigargin 95%

Lieferant: Thermo Fisher Scientific
Gefahr

J62866.MA
J62866.MAEA 843 EUR
J62866.MA
Thapsigargin 95%
Thapsigargin

Cell permeable sesquiterpene lactone tumor promoter. Inhibits endoplasmic reticulum calcium-ATPase


Applications: Cell permeable sesquiterpene lactone tumor promoter. Inhibits endoplasmic reticulum calcium-ATPase

Formel: C₃₄H₅₀O₁₂
MDL: MFCD00083511
CAS-Nummer: 67526-95-8
Merck Index: 14,9272

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR