U-73122 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(6-(((8R,9S,13S,14S,17S)-3-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthren-17-yl)amino)hexyl)-1H-pyrrole-2,5-dione, 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione

J62898.MA
J62898.MAEA 224 EUR
J62898.MA
U-73122 95%
U-73122

Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors.


  • Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml. May require heating or overnight mixing. Insoluble in water


Off-white crystalline solid

Formel: C₂₉H₄₀N₂O₃
Molecular Weight: 464,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00893825
CAS-Nummer: 112648-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR