Destaining bags, ethidium bromide
Lieferant: Thermo Fisher Scientific
J62931.8U
J62931.8UEA 130 EUR
J62931.8U
Destaining bags, ethidium bromide
Elektrophorese-Reagenzien Reagenzien zur Entfernung von Farbstoffen
Ethidium bromide destaining bags are used to safely remove ethidium bromide from nucleotide solutions and gels during overnight treatment. Each bag extracts up to 5 mg of Ethidium Bromide from solution, thus minimising the exposure of research personnel to this material.
  • The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration
  • Provides a safer, more economical means for disposal of hazardous waste
  • Also removes Coomassie stains
Applications: For removal of ethidium bromide from nucleotide solutions and gels. Maximum capacity is 5mg ethidium bromide. Also removes Coomassie stains