Folsäure Reagenz für die Zellkultur
Lieferant: Thermo Fisher Scientific
Synonyme:
Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
J62937.06
J62937.14
J62937.22
J62937.06EA
34.4
EUR
J62937.06
J62937.14
J62937.22
Folsäure Reagenz für die Zellkultur
Folsäure
CAS No.: 59-30-3
Molecular Formula: C19H19N7O6
Formula Weight: 441.41
Physical Form: Powder
Appearance: Yellow to orange
Beilstein Reference: 100781
Merck Reference: 14,4221
MDL No.: MFCD00079305
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells
Formel:
C₁₉H₁₉N₇O₆ Molecular Weight: 441.41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079305 CAS-Nummer: 59-30-3 EINECS: 200-419-0 Merck Index: 14,4221 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...